missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-2/CD22 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1968T-100UG
This item is not returnable.
View return policy
Description
Siglec-2/CD22 Monoclonal specifically detects Siglec-2/CD22 in Human samples. It is validated for Flow Cytometry.
Specifications
| Siglec-2/CD22 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| CAA42006 | |
| CD22 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-2/CD22 Asp20-Arg687 Accession # CAA42006 | |
| RUO | |
| 933 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 219934 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, Siglec2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Siglec-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Siglec-3, -5, -7, -9, -10, -F, or recombinant mouse Siglec-11 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction