missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-11 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3258-SP
This item is not returnable.
View return policy
Description
Siglec-11 Monoclonal specifically detects Siglec-11 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| Siglec-11 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
| Q96RL6 | |
| SIGLEC11 | |
| Human Siglec-11 synthetic peptide PPARLSWTRWGQTVGPSQPSDPGVLELPPIC Accession # Q96RL6 | |
| 25 μg | |
| Primary | |
| Detects human Siglec-11 in direct ELISAs. In direct ELISAs, no cross-reactivity with Siglec-16 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 705904 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| sialic acid binding Ig-like lectin 11, sialic acid-binding Ig-like lectin 11, Sialic acid-binding lectin 11, siglec-11, Siglec11 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 114132 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction