missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-1/CD169 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB5197N-100UG
This item is not returnable.
View return policy
Description
Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Human samples. It is validated for Flow Cytometry.
Specifications
| Siglec-1/CD169 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9BZZ2 | |
| SIGLEC1 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-1/CD169 Ser20-Gln1641 Accession # Q9BZZ2 | |
| RUO | |
| 6614 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 908102 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin, Siglec1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Siglec-1/CD169 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction