missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human sICAM-1 Platinum ELISA Kit, for in vitro diagnostic use
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
Includes
- Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sICAM-1
- HRP-Conjugate anti-human sICAM-1 monoclonal antibody
- Human sICAM-1 Standard, 100ng/mL
- Control high, lyophilized
- Control low, lyophilized
- Sample Diluent
- Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
- Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
- Substrate Solution (tetramethyl-benzidine)
- Stop Solution (1M Phosphoric acid)
- Blue-Dye
- Green-Dye
- Adhesive Films
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
| Assay Sensitivity | 2.2ng/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 96 Tests |
| Synonym | Intercellular adhesion molecule 1, CD54 antigen |
| Target | CD54 (ICAM-1), sCD54 (sICAM-1) |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction