Learn More
Invitrogen™ Human ICAM-1 ELISA Kit, Extra Sensitive
Beschreibung
Beinhaltet
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sICAM-1
HRP-Conjugate anti-human sICAM-1 monoclonal antibody
Human sICAM-1 Standard, 10ng/mL
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films
The Human Intercellular Adhesion Molecule-1 (Hu ICAM-1) ELISA quantitates Hu ICAM-1 in human serum, plasma, amniotic fluid, spontaneous urine. bile, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ICAM-1. Principle of the method The Human ICAM-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Spezifikation
Spezifikation
| Accession Number | P05362 |
| Assay Range | 0.63 to 10.0 ng/mL |
| Assay Sensitivity | 0.33 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Amniotic Fluid, Bile, Plasma, Serum, Supernatant, Urine |
| Target Species | Human |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 3383 |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.