missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SI (aa 36-177) Control Fragment Recombinant Protein

Product Code. 30200355
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200355

Brand: Invitrogen™ RP90016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (61%), Rat (61%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52871. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a sucrase-isomaltase enzyme that is expressed in the intestinal brush border. The encoded protein is synthesized as a precursor protein that is cleaved by pancreatic proteases into two enzymatic subunits sucrase and isomaltase. These two subunits heterodimerize to form the sucrose-isomaltase complex. This complex is essential for the digestion of dietary carbohydrates including starch, sucrose and isomaltose. Mutations in this gene are the cause of congenital sucrase-isomaltase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14410
Concentration 2.20 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 6476
Name Human SI (aa 36-177) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2010204N08Rik; alpha-glucosidase; c-sis; fd42f04; fd44a09; Isomaltase; maltase-glucoamylase (alpha-glucosidase); maltase-glucoamylase, intestinal; mgam; oligosaccharide alpha-1,6-glucosidase; pdgf protein; PDGF subunit B; PDGF-2; Pdgfb; platelet derived growth factor, B polypeptide; platelet-derived growth factor B chain; Platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); platelet-derived growth factor subunit B; Platelet-derived growth factor, same as Sis (Simian sarcoma viral oncogene homologue, c-sis); pro-sucrase-isomaltase (EC 3.2.1.48-10); SI; SIS; Si-s; SUCIMAL; Sucrase; sucrase isomaltase (alpha-glucosidase); sucrase isomaltase, structural; sucrase-isomaltase; sucrase-isomaltase (alpha-glucosidase); sucrase-isomaltase, intestinal; wu:fd42f04; wu:fd44a09
Common Name SI
Gene Symbol SI
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PAVDEISDSTSTPATTRVTTNPSDSGKCPNVLNDPVNVRINCIPEQFPTEGICAQRGCCWRPWNDSLIPWCFFVDNHGYNVQDMTTTSIGVEAKLNRIPSPTLFGNDINSVLFTTQNQTPNRFRFKITDPNNRRYEVPHQYV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.