missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SHOX (aa 50-105) Control Fragment Recombinant Protein

Product Code. 30201389
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30201389 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30201389 Supplier Invitrogen™ Supplier No. RP107034

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (27%), Rat (27%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66351 (PA5-66351. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Homeodomain proteins (HP) are transcriptional regulators that coordinate the expression of genes involved in development, differentiation and cellular transformation. HPs are characterized by a conserved domain of 60 amino acid residues that recognize and bind a site in the regulatory region of the target gene. SHOX, a member of the bicoid subfamily of the paired homeobox family, controls fundamental aspects of growth and development and undergoes splicing resulting in two isoforms, SHOXA and SHOXB. SHOXA is expressed in skeletal muscle, placental, pancreas, heart and bone marrow fibroblast and, to a lesser extent, in kidney and lung. SHOXB is highly expressed in osteogenic cells, but not expressed in brain, kidney, liver or lung. Defects in the gene SHOXX cause Leri-Weill dyschondrosteosis (LWD) and Langer mesomelic dysplasia (LMD).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15266
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6473
Name Human SHOX (aa 50-105) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias GCFX; growth control factor, X-linked; PHOG; Pseudoautosomal homeobox-containing osteogenic protein; short stature homeobox; short stature homeobox protein; Short stature homeobox-containing protein; SHOX; SHOXY; SS
Common Name SHOX
Gene Symbol SHOX
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GTSDSSLQDITEGGGHCPVHLFKDHVDNDKEKLKEFGTARVAEGIYECKEKREDVK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.