missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SHIP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5389-SP
This item is not returnable.
View return policy
Description
SHIP2/INPPL1 Monoclonal specifically detects SHIP2/INPPL1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| SHIP2/INPPL1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| O15357 | |
| INPPL1 | |
| E. coli-derived recombinant human SHIP2 Ala1106-Lys1258 Accession # O15357 | |
| 25 μg | |
| Primary | |
| Detects human SHIP2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse SHIP2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 737935 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.3, EC 3.1.3.n1, inositol polyphosphate phosphatase-like 1, Inositol polyphosphate phosphatase-like protein 1, INPPL1, INPPL-1, phosphatidylinositol-34,5-trisphosphate 5-phosphatase 2, Protein 51C, SH2 domain-containing inositol phosphatase 2, SH2 domain-containing inositol-5'-phosphatase 2, SHIP-2, SHIP251C protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3636 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction