missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SH3TC2 (aa 315-396) Control Fragment Recombinant Protein

Product Code. 30197228
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197228

Brand: Invitrogen™ RP102781

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58058 (PA5-58058. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein with two N-terminal Src homology 3 (SH3) domains and 10 tetratricopeptide repeat (TPR) motifs, and is a member of a small gene family. The gene product has been proposed to be an adapter or docking molecule. Mutations in this gene result in autosomal recessive Charcot-Marie-Tooth disease type 4C, a childhood-onset neurodegenerative disease characterized by demyelination of motor and sensory neurons.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TF17
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79628
Name Human SH3TC2 (aa 315-396) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CMT4C; D430044G18Rik; KIAA1985; MNMN; PP12494; RGD1309038; SH3 domain and tetratricopeptide repeat-containing protein 2; SH3 domain and tetratricopeptide repeats 2; SH3 domain and tetratricopeptide repeats-containing protein 2; SH3TC2
Common Name SH3TC2
Gene Symbol SH3TC2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SGQVGFVPTRNIDPDSYSPMSRNSAFLSDEERCSLLALGSDKQTECSSFLHTLARTDITSVYRLSGFESIQNPPNDLSASQP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado