missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SH3GLB2 (aa 165-246) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54407 (PA5-54407. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
SH3GLB2 (SH3 Domain Containing GRB2 Like, Endophilin B2) is a Protein Coding gene. Among its related pathways are Endocytosis. An important paralog of this gene is SH3GLB1. [GeneCards]
Specifications
Specifications
| Accession Number | Q9NR46 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 56904 |
| Name | Human SH3GLB2 (aa 165-246) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | EMBL:AAH87652.1}; endophilin-B2; Kiaa1848; PP578; PP6569; PP9455; retinoid receptor-induced gene 1; RRIG1; SH3 domain containing GRB2 like endophilin B2; SH3 domain-containing GRB2-like protein B2; SH3-containing protein SH3GLB2; SH3-domain GRB2-like endophilin B2; Sh3glb2; sh3glb2 {ECO:0000312; SPAS-1; stimulator of prostatic adenocarcinoma-specific T-cells-1 |
| Common Name | SH3GLB2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion