missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SH3BGRL3 (aa 22-93) Control Fragment Recombinant Protein

Product Code. 30198279
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198279

Brand: Invitrogen™ RP94257

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56575 (PA5-56575. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SH3BGRL3 could act as a modulator of glutaredoxin biological activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H299
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 83442
Name Human SH3BGRL3 (aa 22-93) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110004L05Rik; AU022188; epididymis secretory protein Li 297; HEL-S-297; P1725; SH3 domain binding glutamate rich protein like 3; SH3 domain binding glutamate-rich protein like 3; SH3 domain binding glutamic acid-rich protein like 3; SH3 domain binding glutamic acid-rich protein-like 3; SH3 domain-binding glutamic acid-rich-like protein 3; SH3 domain-binding protein 1; SH3BGRL3; SH3BGRL3-like protein; SH3BP-1; TIP-B1; TNF inhibitory protein
Common Name SH3BGRL3
Gene Symbol SH3BGRL3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SEVTRILDGKRIQYQLVDISQDNALRDEMRALAGNPKATPPQIVNGDQYCGDYELFVEAVEQNTLQEFLKLA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato