missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SGCB (aa 241-318) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52835 (PA5-52835. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function of this protein remains unknown.
Specifications
Specifications
| Accession Number | Q16585 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 6443 |
| Name | Human SGCB (aa 241-318) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 43 kDa dystrophin-associated glycoprotein; 43 DAG; 43 kDa; A3b; AI747103; AI844814; beta sarcoglycan; beta-sarcoglycan; beta-sarcoglycan(43 kD dystrophin-associated glycoprotein); beta-SG; fa03a05; LGMD2E; limb girdle muscular dystrophy 2 E (non-linked families); sarcoglycan beta; sarcoglycan, beta (43 kD dystrophin-associated glycoprotein); sarcoglycan, beta (43 kDa dystrophin-associated glycoprotein); sarcoglycan, beta (dystrophin-associated glycoprotein); SGC; Sgcb; wu:fa03a05; zgc:110833 |
| Common Name | SGCB |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction