missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SFXN3 Partial ORF (AAH00124.2, 125 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00081855-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16075695

  • £429.00 / 25µg
Estimated Shipment: 09-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: WVNQSFNAIVNYSNRSGDTPITVRQLGTAYVSATTGAVATALGLKSLTKHLPPLVGRFVPFAAVAAANCINIPLMRQRELQVGIPVADEAGQRLGYSVTA
Specifications

Specifications

AAH00124.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
WVNQSFNAIVNYSNRSGDTPITVRQLGTAYVSATTGAVATALGLKSLTKHLPPLVGRFVPFAAVAAANCINIPLMRQRELQVGIPVADEAGQRLGYSVTA
RUO
SFXN3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
81855
SFXN3 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BA108L7.2/SFX3
SFXN3
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers