missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human sFRP-4 (NP_003005) VersaClone cDNA
Description
- Also known as: PYL; FRP-4; FRPHE; FRZB-2; sFRP-4
- SFRP4 is a member of the secreted frizzled-related protein family that contains a cysteine-rich domain homologous to the putative Wntbinding site of Frizzled proteins.
- SFRPs act as soluble modulators of Wnt signaling.
- The expression of SFRP4 in ventricular myocardium correlates with apoptosis related gene expression.
- The sFRP-4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hSFRP4 |
| Accession Number | NP_003005 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1054 bp |
| Gene ID (Entrez) | 6424 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?