missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human sFRP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6838-SP
This item is not returnable.
View return policy
Description
sFRP-2 Monoclonal specifically detects sFRP-2 in Human samples. It is validated for Western Blot.
Specifications
| sFRP-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q96HF1 | |
| SFRP2 | |
| Mouse myeloma cell line NS0-derived recombinant human sFRP-2 Leu25-Cys295 Accession # Q96HF1 | |
| 25 μg | |
| Primary | |
| Detects human sFRP-2 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) sFRP-2 is observed and no cross-reactivity with recombinant human (rh) sFRP-1 or rhsFRP-5 is observed. In Western blots, no cross-reactivity with rhsFRP-1, -4, -5, or rmsFRP-2 is observed. 100% cross-reactivity with rhsFRP-3 is observed under non-reducing conditions only. | |
| Human | |
| Purified |
| Western Blot | |
| 755644 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2, sFRP2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6423 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction