missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SF3B2 (aa 808-894) Control Fragment Recombinant Protein

Product Code. 30195388
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195388

Brand: Invitrogen™ RP100756

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60902 (PA5-60902. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Subunit of the splicing factor SF3B required for 'A' complex assembly formed by the stable binding of U2 snRNP to the branchpoint sequence (BPS) in pre-mRNA. Sequence independent binding of SF3A/SF3B complex upstream of the branch site is essential, it may anchor U2 snRNP to the pre-mRNA. May also be involved in the assembly of the 'E' complex. Belongs also to the minor U12-dependent spliceosome, which is involved in the splicing of rare class of nuclear pre-mRNA intron.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13435
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10992
Name Human SF3B2 (aa 808-894) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 145 kDa; 2610311M13Rik; 2810441F20Rik; B230398H18Rik; Cus1; pre-mRNA splicing factor SF3b 145 kDa subunit; pre-mRNA-splicing factor SF3b 145 kDa subunit; SAP 145; SAP145; SF3b1; SF3B145; SF3b150; SF3B2; spliceosome associated protein 145; spliceosome-associated protein 145; Splicing factor 3 B subunit 2; splicing factor 3 b, subunit 2; splicing factor 3 b, subunit 2, 145 kD; splicing factor 3 b, subunit 2, 145 kDa
Common Name SF3B2
Gene Symbol SF3B2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MSTVMSRKGPAPELQGVEVALAPEELELDPMAMTQKYEEHVREQQAQVEKEDFSDMVAEHAAKQKQKKRKAQPQDSRGGSKKYKEFK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.