missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SF3B1 (aa 33-117) Control Fragment Recombinant Protein

Product Code. 30205002
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205002

Brand: Invitrogen™ RP101202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83997 (PA5-83997. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes subunit 1 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex. The splicing factor 3b/3a complex binds pre-mRNA upstream of the intron's branch site in a sequence independent manner and may anchor the U2 snRNP to the pre-mRNA. Splicing factor 3b is also a component of the minor U12-type spliceosome. The carboxy-terminal two-thirds of subunit 1 have 22 non-identical, tandem HEAT repeats that form rod-like, helical structures. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75533
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23451
Name Human SF3B1 (aa 33-117) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 155 kDa; 2810001M05Rik; AA409119; Hsh155; LOW QUALITY PROTEIN: splicing factor 3 B subunit 1; MDS; pre-mRNA processing 10; pre-mRNA splicing factor SF3b, 155 kDa subunit; pre-mRNA-splicing factor SF3b 155 kDa subunit; PRP10; PRPF10; SAP 155; SAP155; SF3B1; SF3b155; spliceosome-associated protein 155; Splicing factor 3 B subunit 1; splicing factor 3 b subunit 1, 155 kDa; splicing factor 3 b, subunit 1; splicing factor 3 b, subunit 1, 155 kDa; splicing factor 3 b, subunit 1, 155 kD; splicing factor 3 b, subunit 1, 155 kDa; TA-8; Targ4; transforming growth factor alpha regulated gene 4; tst; wu:fb99f09
Common Name SF3B1
Gene Symbol SF3B1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LDSTGYYDQEIYGGSDSRFAGYVTSIAATELEDDDDDYSSSTSLLGQKKPGYHAPVALLNDIPQSTEQYDPFAEHRPPKIADRED
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.