missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SF20/MYDGF Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC1147R-100UG
This item is not returnable.
View return policy
Description
SF20/MYDGF Monoclonal specifically detects SF20/MYDGF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| SF20/MYDGF | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL-25, IL25IL27w, IL27, IL-27, interleukin 25, interleukin 27 working designation, Interleukin-25, Ly6elg, MYDGF, R33729_1, SF20hypothetical protein LOC56005, stromal cell-derived growth factor, Stromal cell-derived growth factor SF20 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human SF20 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 1009420 | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| Q969H8 | |
| MYDGF | |
| Chinese hamster ovary cell line CHO-derived recombinant human SF20, Met1-Leu173, Accession # Q969H8 | |
| 100 μg | |
| 56005 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction