missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SETDB2 Partial ORF (NP_114121.1, 451 a.a. - 550 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00083852-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16075705

  • £283.00 / 10µg
Estimated Shipment: 08-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Proteins that contain a SET domain, such as SETDB2, modulate gene expression epigenetically through histone H3 (see MIM 601128) methylation. SETDB2 is likely a histone H3 methyltransferase, as it contains both the active site and flanking cysteine residues required for catalytic activity (Zhang et al., 2003 [PubMed 12754510]).[supplied by OMIM]

Sequence: HPRTAKTEKCPPKFSNNPKELTMETKYDNISRIQYHSVIRDPESKTAIFQHNGKKMEFVSSESVTPEDNDGFKPPREHLNSKTKGAQKDSSSNHVDEFED
Specifications

Specifications

NP_114121.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HPRTAKTEKCPPKFSNNPKELTMETKYDNISRIQYHSVIRDPESKTAIFQHNGKKMEFVSSESVTPEDNDGFKPPREHLNSKTKGAQKDSSSNHVDEFED
RUO
SETDB2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
83852
SETDB2 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C13orf4/CLLD8/CLLL8/DKFZp586I0123/DKFZp761J1217/KMT1F
SETDB2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers