missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Human Serum, Type AB
Golden Yellow Liquid. Proven to grow many human cell lines at a faster rate and with a smaller percentage of serum than mixed blood group serum.
Brand: MP Biomedicals™ 092938249
Code : NEW
Additional Details : Weight : 0.12780kg
Specifications
U.S. regulations | |
-20°C | |
Liquid | |
100 mL | |
0.1 μm filtration | |
Yes | |
No | |
7 to 8 |
Golden Yellow | |
Human | |
Human Serum | |
Sterile | |
Grow many Human cell lines at a faster rate | |
≤26mg/dL | |
263 to 323mOsm/kg | |
5.5 to 7.5 g/dL |