missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Amyloid A4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3856-SP
This item is not returnable.
View return policy
Description
Serum Amyloid A4 Polyclonal specifically detects Serum Amyloid A4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Serum Amyloid A4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-SAA, C-SAACSAAConstitutively expressed serum amyloid A protein, SAA4, SAA5, serum amyloid A-4 protein, serum amyloid A4, constitutive | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6291 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P35542 | |
| SAA4 | |
| E. coli-derived recombinant human Serum Amyloid A4 Glu19-Tyr130 Accession # P35542 | |
| 25 μg | |
| Primary | |
| Detects human Serum Amyloid A4 in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse Serum Amyloid A4 is observed and less than 5% cross-reactivity with recombinant human (rh) Serum Amyloid A1 and rhSerum Amyloid A2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction