missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Amyloid A1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3019-SP
This item is not returnable.
View return policy
Description
Serum Amyloid A1 Monoclonal specifically detects Serum Amyloid A1 in Human samples. It is validated for Western Blot.
Specifications
| Serum Amyloid A1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P02735 | |
| SAA1 | |
| E. coli-derived recombinant human SAA1 Arg19-Tyr122 Accession # P02735 | |
| 25 μg | |
| Primary | |
| Detects human Serum Amyloid A1 (SAA1) in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse SAA1 and no cross-reactivity with recombinant human (rh) SAA4 is observed. In direct ELISAs, no cross-reactivity with rhSAA2 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 370820 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC111216, PIG4, SAA, SAA1, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6288 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction