missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Serum Albumin Polyclonal Antibody
GREENER_CHOICE

Chicken Polyclonal Antibody

Brand:  Invitrogen™ PA172054

Product Code. 11568301

  • £190.00 / 10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PA1-72054 detects human serum albumin in human samples. PA1-72054 has been successfully used in ELISA and western blot procedures. The PA1-72054 immunogen is from purified human serum albumin.

Albumin is a soluble, monomeric protein with a molecular weight of approximately 65-67 kDa that comprises about one-half of the blood serum protein. Human serum albumin (HAS) is the most abundant protein in human blood plasma, and has a serum half-life of approximately 20 days. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular, unglycosylated serum protein synthesized in the liver as pre-pro-albumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough 'produce the secreted albumin. Serum albumin has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. The main function of albumin is the regulation of the colloidal osmotic pressure of blood.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Human Serum Albumin
Polyclonal
Unconjugated
ALB
Alb; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341
Chicken
IgY fraction
RUO
213
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
10 mg/mL
PBS with 0.075% sodium azide; pH 7.2
P02768
ALB
Purified human serum albumin.
10 mg
Primary
Human
Antibody
IgY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.