missing translation for 'onlineSavingsMsg'
Learn More

Human Serum Albumin Antibody (10C7), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-78591-0.025mg

 View more versions of this product

Product Code. 18793374

  • £149.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Human Serum Albumin Monoclonal specifically detects Human Serum Albumin in Human samples. It is validated for ELISA, Immunoprecipitation, Radioimmunoassay.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Human Serum Albumin
Monoclonal
1 mg/mL
ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Mouse
Affinity Purified
RUO
Primary
Human
IgG2a
ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
10C7
Unconjugated
P02768
ALB
Human Serum Albumin
0.025 mg
Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
213
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only