missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Serpin G1/C1 Inhibitor Quantikine ELISA Kit
Specifications
Specifications
| Assay Range | 62.50 to 2000 pg/mL |
| Assay Sensitivity | 25.6 pg/mL |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Saliva, Urine |
| Specificity | Natural human Serpin G1. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 710 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | C1 Inhibitor, C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1 |
| Target | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?