missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin G1/C1 Inhibitor Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2488-SP
This item is not returnable.
View return policy
Description
Serpin G1/C1 Inhibitor Polyclonal specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.
Specifications
| Serpin G1/C1 Inhibitor | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SERPING1 | |
| Human plasma-derived Serpin G1/C1 Inhibitor | |
| 25 μg | |
| Primary | |
| Detects human Serpin G1/C1 Inhibitor in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin A5, rhSerpin C1, rhSerpin F1, rhSerpin F2, rhSerpin I1, rhSerpin I2, recombinant mouse (rm) Serpin D1 and rmSerpin E2 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| C1 Inhibitor, C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 710 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction