missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin F2/alpha 2-Antiplasmin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1470
This item is not returnable.
View return policy
Description
Serpin F2/alpha 2-Antiplasmin Polyclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.
Specifications
| Serpin F2/alpha 2-Antiplasmin | |
| Polyclonal | |
| Unconjugated | |
| P08697 | |
| SERPINF2 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin F2/alpha 2 -Antiplasmin Met28-Lys491 Accession # P08697 | |
| 100 μg | |
| Primary | |
| Detects human Serpin F2/alpha 2 -Antiplasmin in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse (rm) Serpin F2 is observed and less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin A5, rmSerpin C1, and rmSerpin D1 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 4.2 ug/mL | |
| AAPclade F (alpha-2 antiplasmin, alpha 2-Antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5345 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction