missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin E2/PN1 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF2980
This item is not returnable.
View return policy
Description
Serpin E2/PN1 Polyclonal specifically detects Serpin E2/PN1 in Human samples. It is validated for Western Blot.
Specifications
| Serpin E2/PN1 | |
| Polyclonal | |
| Biotin | |
| NP_001130000 | |
| SERPINE2 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin E2/PN1 Met1-Pro397 Accession # NP_001130000 | |
| 50 μg | |
| Primary | |
| Detects human Serpin E2/PN1 in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse (rm) Serpin E2 is observed and less than 1% cross-reactivity with recombinant human (rh) Serpin E1, rhSerpin I1, rhSerpin I2, and rhSerpin B6 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| GDN, GdNPF, GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Proteinase Nexin 1, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5270 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction