Learn More
Human Serpin E2/PN1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand:  R&D Systems AF2980
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18049971
  • £464.80 / 100µg
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
Serpin E2/PN1 Polyclonal specifically detects Serpin E2/PN1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Serpin E2/PN1
Polyclonal
Unconjugated
NP_001130000
SERPINE2
Mouse myeloma cell line NS0-derived recombinant human Serpin E2 Met1-Pro397 Accession # NP_001130000
100 μg
Primary
Detects human Serpin E2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse Serpin E2 is observed and less than 1% cross-reactivity with recombinant human (rh) Serpin E1, rhSerpin I1, rhSerpin I2, and rhSerpin B6 is observed.
Human
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
GDN, GdNPF, GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Proteinase Nexin 1, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2
Goat
Affinity Purified
RUO
5270
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.