missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Serpin E1/PAI-1 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Pack (6 Plates)
One 96-well plate
Unit Size:
1 set
Pack of 96
Description
The Human Serpin E1/PAI-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Serpin E1/PAI-1 in cell culture supernates (50 ul), cell lysates (50 ul), edta plasma (50 ul), heparin plasma (50 ul), citrate plasma (50 ul), ctad plasma (50 ul).
Specifications
Specifications
| Assay Range | 0.31 to 20 ng/mL |
| Assay Sensitivity | 0.142 ng/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Citrate, Heparin, CTAD) |
| Specificity | Natural and Recombinant Human Serpin E1 |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 5054 |
| Quantity | 1 Pack (6 Plates) |
| Sample Volume | Cell Culture Supernates (50 uL),Cell Lysates (50 uL),EDTA Plasma (50 uL),Heparin Plasma (50 uL),Citrate Plasma (50 uL),CTAD Plasma (50 uL) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction