missing translation for 'onlineSavingsMsg'
Learn More

Human Serpin E1/PAI-1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 4 publications

£181.00 - £1765.00

Specifications

Antigen Serpin E1/PAI-1
Clone 242816
Concentration LYOPH
Dilution Western Blot 1 ug/mL, Simple Western 2.5 ug/mL, Neutralization 0.3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, Neutralization, ELISA
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18785539
GREENER_CHOICE
View Documents
R&D Systems
MAB1786-SP
25 μg
£181.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18702558
GREENER_CHOICE
View Documents
R&D Systems
MAB1786-100
100 μg
£447.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18762548
GREENER_CHOICE
View Documents
R&D Systems
MAB1786-500
500 μg
£1765.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Serpin E1/PAI-1 Monoclonal specifically detects Serpin E1/PAI-1 in Human samples. It is validated for Western Blot, Simple Western, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

Serpin E1/PAI-1
LYOPH
Western Blot, Neutralization, ELISA
Unconjugated
Mouse
Human
P05121
5054
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin E1/PAI-1 Met1-Pro402 Accession # P05121
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Serpin E1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Serpin A1, A3, A4, A8, C1, F1, F2, I1, I2, recombinant mouse Serpin D1 or E2 is observed.
242816
Western Blot 1 ug/mL, Simple Western 2.5 ug/mL, Neutralization 0.3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Endothelial plasminogen activator inhibitor, Nexin, PAI1, PAI-1, PAI1PAI-1, PAISerpin E1, PLANH1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1
SERPINE1
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.