missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin D1/Heparin Cofactor II Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3198-SP
This item is not returnable.
View return policy
Description
Serpin D1/Heparin Cofactor II Monoclonal specifically detects Serpin D1/Heparin Cofactor II in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Serpin D1/Heparin Cofactor II | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P05546 | |
| SERPIND1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin D1/Heparin Cofactor II Gly20-Arg498 Accession # P05546 | |
| 25 μg | |
| Primary | |
| Detects human Serpin D1/Heparin Cofactor II in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Serpin A3, A4, B6, B8, C1, I1, I2, recombinant mouse Serpin A1, A6, D1, E1, E2, F1, or F2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 373008 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HC2, HC2LS2, HCF2, HCF2clade D (heparin cofactor), member 1, HC-II, Heparin cofactor II, HLS2HCII, leuserpin 2, LS2, Protease inhibitor leuserpin-2, Serpin D1, serpin peptidase inhibitor, clade D (heparin cofactor), member 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3053 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction