missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin B8/Proteinase Inhibitor 8 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB4158
This item is not returnable.
View return policy
Description
Serpin B8/Proteinase Inhibitor 8 Monoclonal specifically detects Serpin B8/Proteinase Inhibitor 8 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Serpin B8/Proteinase Inhibitor 8 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P50452 | |
| SERPINB8 | |
| S. frugiperda insect ovarian cell line Sf21-derived recombinant human Serpin B8/Proteinase Inhibitor 8 Asp2-Pro374 Accession # P50452 | |
| 100 μg | |
| Primary | |
| Detects human Serpin B8/Proteinase Inhibitor 8 in direct ELISAs. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse (rm) Serpin B8 is observed. No cross-reactivity with recombinant human Serpin A1, A3, A4, A5, A6, A9, A11, A12, B3, B6, B9, C1, D1, E1, E2, F1, F2, G1, I1, I2, rmSerpin A1, A3N, A6, A9, C1, D1, E1, E2, F1, F2, I1, recombinant rat (rr) Serpin A6, or rrSerpin A12 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 423023 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAP2, CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, NK10, Peptidase inhibitor 8, PI8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5271 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction