missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin B2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8550
This item is not returnable.
View return policy
Description
SerpinB2 Monoclonal specifically detects SerpinB2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated.
Specifications
| SerpinB2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HsT1201, Monocyte Arg-serpin, PAI, PAI2, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2, Urokinase Inhibitor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5055 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, KnockDown | |
| 930109 | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready, Knockout Validated | |
| P05120 | |
| SERPINB2 | |
| E. coli-derived recombinant human Serpin B2 Glu2-Ala65, Ala99-Pro415 Accession # P05120 | |
| 100 μg | |
| Primary | |
| Detects human Serpin B2 in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction