missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin A9/Centerin Antibody, R&D Systems™
Mouse Monoclonal Antibody
£359.60
Specifications
Antigen | Serpin A9/Centerin |
---|---|
Clone | 585302 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Description
Serpin A9/Centerin Monoclonal specifically detects Serpin A9/Centerin in Human samples. It is validated for Immunohistochemistry.Specifications
Serpin A9/Centerin | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
AAQ89063 | |
327657 | |
Mouse myeloma cell line NS0-derived recombinant human Serpin A9/Centerin Ala24-Thr417 Accession # AAQ89063 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Serpin A9/Centerin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Serpin A1, A3, A4, A5, A6, A11, A12, B6, B8, B9, C1, D1, E1, E2, F1, F2, G1, I1, I2, recombinant mouse (rm) SerpinA3N, and rmSerpinA9 is observed. |
585302 | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Centerin, GCET1, SERPINA11, SERPINA11b | |
SERPINA9 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only