missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin A8/Angiotensinogen Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF3156-SP
This item is not returnable.
View return policy
Description
Serpin A8/Angiotensinogen Polyclonal specifically detects Serpin A8/Angiotensinogen in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Serpin A8/Angiotensinogen | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGT, alpha-1 antiproteinase, antitrypsin, angiotensin II, Angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, Aogen, FLJ92595, FLJ97926, PAT, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 183 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 2-15 ug/mL | |
| ABM85122 | |
| AGT | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin A8/Angiotensinogen Asp34-Ala485 Accession # ABM85122 | |
| 25 μg | |
| Primary | |
| Detects human Serpin A8/Angiotensinogen in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse Serpin A8/Angiotensinogen is observed, and less than 5% cross-reactivity with recombinant human Serpin A1, A4, A5, F2, G1, I1, and I2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction