missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Serpin A5/Protein C Inhibitor ELISA Kit

Product Code. 17105199
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17105199

Brand: Invitrogen™ EH412RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Serpin A5/Protein C Inhibitor quantitates human Serpin A5/Protein C Inhibitor in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Serpin A5/Protein C Inhibitor.

This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 4933415L04; Acrosomal serine protease inhibitor; alpha-1 antiproteinase; antitr; antitrypsin; PAI3; PAI-3; Pci; PCI-B; PLANH3; Plasma serine protease inhibitor; Plasminogen activator inhibitor 3; plasminogen activator inhibitor III; plasminogen activator inhibitor-3; PROCI; Protein C inhibitor; serine (or cysteine) peptidase inhibitor, clade A, member 5; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serine (or cysteine) proteinase inhibitor, clade A, member 5; serpin A5; serpin family A member 5; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serpin peptidase inhibitor, clade A, member 5; Serpina5
Assay Range 24.58 to 6,000 pg/mL
Assay Sensitivity 24 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5104
Gene Symbol SERPINA5
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target SERPINA5
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.