missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin A5/Protein C Inhibitor Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1266
Additional Details : Weight : 0.00970kg
Description
Serpin A5/Protein C Inhibitor Monoclonal specifically detects Serpin A5/Protein C Inhibitor in Human samples. It is validated for Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
Serpin A5/Protein C Inhibitor | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
P05154 | |
SERPINA5 | |
Mouse myeloma cell line NS0-derived recombinant human Serpin A5/Protein C His20-Pro406 Accession # P05154 | |
500 μg | |
Primary | |
Detects human Serpin A5/Protein C in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Serpin A1, A3, F2, C1, or recombinant mouse Serpin A5 is observed. | |
Human | |
Purified |
Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF | |
210326 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Acrosomal serine protease inhibitor, antitrypsin), member 5, member 5, PAI-3, PAI3PLANH3, PCI, PCIplasminogen activator inhibitor III, Plasminogen activator inhibitor 3, plasminogen activator inhibitor-3, PROCIplasma serine protease inhibitor, Protein C inhibitor, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A5, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), SERPNA5 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5104 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
For Research Use Only