missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin A12 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4410-SP
This item is not returnable.
View return policy
Description
Serpin A12 Monoclonal specifically detects Serpin A12 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Serpin A12 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q8IW75 | |
| SERPINA12 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin A12 Leu20-Lys414 Accession # Q8IW75 | |
| 25 μg | |
| Primary | |
| Detects human Serpin A12 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Serpin A1, A3, A4, A5, A6, B8, 9, C1, D1, E1, F2, G1, I1, I2, recombinant mouse (rm) Serpin E2, F1, recombinant rat Serpin B6, rhA1 Antitrypsin, rhAngiotensinogin, or rmPEDF is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 484223 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antitrypsin), member 12, member 12, OL-64, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 145264 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction