missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin A1/alpha 1-Antitrypsin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB1268-500
This item is not returnable.
View return policy
Description
Serpin A1/alpha 1-Antitrypsin Monoclonal specifically detects Serpin A1/alpha 1-Antitrypsin in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Immunocytochemistry.
Specifications
| Serpin A1/alpha 1-Antitrypsin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P01009 | |
| SERPINA1 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin A1/alpha 1-Antitrypsin Glu25-Lys418 Accession # P01009 | |
| 500 μg | |
| Primary | |
| Detects human Serpin A1/alpha 1-Antitrypsin in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Serpin A3, A4, A5, C1, F1, recombinant mouse Serpin D1, or F2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| 202808 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| A1A, A1AT, AATMGC23330, alpha 1-Antitrypsin, alpha 1-Proteinase Inhibitor, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5265 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction