missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Serine racemase (aa 173-272) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64916 (PA5-64916. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
Specifications
Specifications
| Accession Number | Q9GZT4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 63826 |
| Name | Human Serine racemase (aa 173-272) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs |
| Common Name | Serine racemase |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction