missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Semaphorin 4A Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4694-SP
This item is not returnable.
View return policy
Description
Semaphorin 4A Polyclonal specifically detects Semaphorin 4A in Human samples. It is validated for Western Blot.
Specifications
| Semaphorin 4A | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CORD10, RP35, Sema B, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4A, SEMA4A, SEMAB, semaphorin-4A, Semaphorin-B, SemB, SEMBFLJ12287 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 64218 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q9H3S1 | |
| SEMA4A | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 4A Gly32-His683 Accession # Q9H3S1 | |
| 25 μg | |
| Primary | |
| Detects human Semaphorin 4A in direct ELISAs and Western blots. In direct ELISAs, approximately 80% cross-reactivity with recombinant mouse Semaphorin 4A is observed, and less than 1% cross-reactivity with recombinant human (rh) Semaphorin 4B, rhSemaphorin 4C, and rhSemaphorin 4D is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction