missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Semaphorin 3A Fc Recombinant Protein, PeproTech®

Product Code. 17890683
Click to view available options
Quantity:
10 x 100 μg
100 μg
25 μg
5 μg
5 x 100 μg
Unit Size:
100µg
1mg
25µg
500µg
5µg
Pack of 5
This item is not returnable. View return policy

Product Code. 17890683

Brand: Gibco™ 15017H1MG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

150-17H-1MG will be provided as 10 x 100 μg (150-17H-100UG). 150-17H-500UG will be provided as 5 x 100 μg (150-17H-100UG). Recombinant Human Semaphorin 3A Fc has a calculated molecular weight of 226.2 kDa and therefore runs above the 200kDa marker by SDS-PAGE analysis under nonreducing conditions. When run under reducing conditions, this protein migrates as three distinct bands that, due to glycosylation, run higher than expected at apparent molecular weights of approximately 120-130 kDa, 90-100 kDa, and 35-40 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is a member of the semaphorin family and encodes a protein with an Ig-like C2-type domain, a PSI domain and a Sema domain. This secreted protein can function as either a chemorepulsive agent, inhibiting axonal outgrowth, or as a chemoattractive agent, stimulating the growth of apical dendrites. In both cases, the protein is vital for normal neuronal pattern development. Increased expression of this protein is associated with schizophrenia and is seen in a variety of human tumor cell lines. Also, aberrant release of this protein is associated with the progression of Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14563
For Use With (Application) ELISA, Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 10371
Molecular Weight (g/mol) 226.2
Name Human Semaphorin 3 A Fc
Quantity 10 x 100 μg
Source CHO cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias COLL1; coll-1; collapsin 1; collapsin-1; HH16; Hsema-I; Hsema-III; MGC133243; sema D; sema domain, immunoglobulin domain (Ig), short basic domain, sec; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3 A; sema III; SEMA1; SEMA3A; Semad; SEMAIII; SEMAL; Semaphorin 3 A; semaphorin D; semaphorin III; semaphorin L; semaphorin-3 A; Semaphorin-D; SemD
Gene Symbol SEMA3A
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Sequence HHHHHHHHGK NNVPRLKLSY KEMLESNNVI TFNGLANSSS YHTFLLDEER SRLYVGAKDH IFSFDLVNIK DFQKIVWPVS YTRRDECKWA GKDILKECAN FIKVLKAYNQ THLYACGTGA FHPICTYIEI GHHPEDNIFK LENSHFENGR GKSPYDPKLL TASLLIDGEL YSGTAADFMG RDFAIFRTLG HHHPIRTEQH DSRWLNDPKF ISAHLISESD NPEDDKVYFF FRENAIDGEH SGKATHARIG QICKNDFGGH RSLVNKWTTF LKARLICSVP GPNGIDTHFD ELQDVFLMNF KDPKNPVVYG VFTTSSNIFK GSAVCMYSMS DVRRVFLGPY AHRDGPNYQW VPYQGRVPYP RPGTCPSKTF GGFDSTKDLP DDVITFARSH PAMYNPVFPM NNRPIVIKTD VNYQFTQIVV DRVDAEDGQY DVMFIGTDVG TVLKVVSIPK ETWYDLEEVL LEEMTVFREP TAISAMELST KQQQLYIGST AGVAQLPLHR CDIYGKACAE CCLARDPYCA WDGSACSRYF PTAKRATRAQ DIRNGDPLTH CSDLHHDNHH GHSPEERIIY GVENSSTFLE CSPKSQRALV YWQFQRRNEE RKEEIRVDDH IIRTDQGLLL RSLQQKDSGN YLCHAVEHGF IQTLLKVTLE VIDTEHLEEL LHKDDDGDGS KTKEMSNSMT PSQKVWYRDF MQLINHPNLN TMDEFCEQVW KRDRKQRRQR PGHTPGNSNK WKHLQENKKG RNRRTHEFER APRSVGGPKS CDKTHTCPPC PAPELLGGPS VFLFPPKPKD TLMISRTPEV TCVVVDVSHE DPEVKFNWYV DGVEVHNAKT KPREEQYNST YRVVSVLTVL HQDWLNGKEY KCKVSNKALP APIEKTISKA KGQPREPQVY TLPPSRDELT KNQVSLTCLV KGFYPSDIAV EWESNGQPEN NYKTTPPVLD SDGSFFLYSK LTVDKSRWQQ GNVFSCSVMH EALHNHYTQK SLSLSPGK
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.