missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Seli (aa 68-89) Control Fragment Recombinant Protein

Product Code. 30210191
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210191

Brand: Invitrogen™ RP109057

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139972 (PA5-139972. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The multi-pass transmembrane protein encoded by this gene belongs to the CDP-alcohol phosphatidyltransferase class-I family. It catalyzes the transfer of phosphoethanolamine from CDP-ethanolamine to diacylglycerol to produce phosphatidylethanolamine, which is involved in the formation and maintenance of vesicular membranes, regulation of lipid metabolism, and protein folding. This protein is a selenoprotein, containing the rare selenocysteine (Sec) amino acid at its active site. Sec is encoded by the UGA codon, which normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, designated the Sec insertion sequence (SECIS) element, that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. A pseudogene of this locus has been identified on chromosome 7.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9C0D9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 85465
Name Human Seli (aa 68-89) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias EPT1; ethanolaminephosphotransferase 1; hEPT1; KIAA1724; RGD1560938; SELENOI; Selenoprotein I; SELI; SEPI
Common Name Seli
Gene Symbol SELENOI
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MAYFDPDFYASAPGHKHVPDWV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.