missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SEC23B (aa 172-268) Control Fragment Recombinant Protein

Product Code. 30210884
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210884

Brand: Invitrogen™ RP102455

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52533 (PA5-52533. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The function of this gene product has been implicated in cargo selection and concentration. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15437
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10483
Name Human SEC23B (aa 172-268) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CDAII; CDA-II; CDAN2; CWS7; HEMPAS; hSec23B; Protein transport protein Sec23B; Sec23 homolog B; Sec23 homolog B (S. cerevisiae); Sec23 homolog B, coat complex II component; Sec23 homolog B, COPII coat complex component; Sec23b; SEC23B (S. cerevisiae); SEC23-like protein B; SEC23-related protein B; transport protein SEC23B
Common Name SEC23B
Gene Symbol SEC23B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MVQVHELSCEGISKSYVFRGTKDLTAKQIQDMLGLTKPAMPMQQARPAQPQEHPFASSRFLQPVHKIDMNLTDLLGELQRDPWPVTQGKRPLRSTGV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.