Learn More
Invitrogen™ Human SEC22C (aa 95-173) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65957 (PA5-65957. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q9BRL7 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 9117 |
| Name | Human SEC22C (aa 95-173) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4932412K21; 5930407I15Rik; C530046H07; SEC22 homolog C, vesicle trafficking protein; SEC22 vesicle trafficking protein homolog C; SEC22 vesicle trafficking protein homolog C (S. cerevisiae); SEC22 vesicle trafficking protein-like 3; SEC22 vesicle trafficking protein-like C; SEC22 vesicle trafficking protein-like protein C; SEC22 vesicle-trafficking protein homolog C; SEC22 vesicle-trafficking protein-like 3; SEC22C; Sec22l3; secretion deficient 22 C; Unknown (protein for MGC:134135); UNQ459/PRO784; Vesicle-trafficking protein SEC22c |
| Common Name | SEC22C |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.