missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SDPR Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
SDPR Synthetic Peptide, PEP-1564, from Invitrogen.
The serum deprivation-response protein (SDPR) is a calcium-independent phospholipid-binding protein whose expression is increased in serum-starved cells. SDPR is a substrate for protein kinase C (PKC) phosphorylation and recruits the polymerase I and transcript release factor (PTRF) to caveolae. Removal of this protein causes caveolae loss and its over-expression results in caveolae deformation and membrane tubulation. Both SDPR and PTRF, as well as the other member of the cavin family PRKCDBP were down regulated in breast cancer cell lines and breast tumor tissue, suggesting that expression of the cavin family proteins could be a useful prognostic indicator of breast cancer progression.
Specifications
Specifications
| Gene ID (Entrez) | 8436 |
| Species | Human |
| Purification Method | purified |
| Accession Number | O95810 |
| Common Name | Human SDPR |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction