missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SCO1 (aa 230-298) Control Fragment Recombinant Protein

Product Code. 30210384
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210384

Brand: Invitrogen™ RP93316

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54452 (PA5-54452. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Synthesis of cytochrome c oxidase 1 was initially identified in yeast as one of two cytochrome c oxidase (COX) assembly proteins that enable the assembly of cytochrome c holoenzyme, a complex that catalyzes the transfer of reducing equivalents from cytochrome c to molecular oxygen and pumps protons across the inner mitochondrial membrane. Like their yeast homologs, the function of both SCO1 and SCO2 are dependent on copper ion binding. Mutations in either gene can lead to cytochrome c oxidase respiratory chain defects, with a missense mutation in human SCO1 (P174L) associated with a fatal neonatal hepatopathy when the second allele is also non-functional, suggesting the pathology is due to loss of function. It has been suggested that this mutation alters the SCO1 affinity for the copper (I) ion, thus impairing the efficiency of copper transfer to the cytochrome c oxidase.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75880
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6341
Name Human SCO1 (aa 230-298) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610001C07Rik; D11Bwg1310e; Protein SCO1 homolog, mitochondrial; RGD1559538; SCO cytochrome c oxidase assembly protein 1; SCO cytochrome oxidase deficient homolog 1; SCO cytochrome oxidase deficient homolog 1 (yeast); SCO1; SCO1 cytochrome c oxidase assembly protein; SCO1, cytochrome c oxidase assembly protein; SCOD1; synthesis of cytochrome C oxidase 1
Common Name SCO1
Gene Symbol SCO1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TREEVDQVARAYRVYYSPGPKDEDEDYIVDHTIIMYLIGPDGEFLDYFGQNKRKGEIAASIATHMRPYR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.