missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SCL/Tal1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3360
This item is not returnable.
View return policy
Description
SCL/Tal1 Polyclonal specifically detects SCL/Tal1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| SCL/Tal1 | |
| Polyclonal | |
| Unconjugated | |
| P17542 | |
| TAL1 | |
| E. coli-derived recombinant human SCL Arg199-Gly266 Accession # P17542 | |
| 100 μg | |
| Primary | |
| Detects human SCL in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, Tal1, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5, TCL5tal-1 product | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6886 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction