missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SCGF/CLEC11a Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF1904
Additional Details : Weight : 0.00970kg
Description
SCGF/CLEC11a Polyclonal specifically detects SCGF/CLEC11a in Human samples. It is validated for Western Blot.Specifications
SCGF/CLEC11a | |
Polyclonal | |
Unconjugated | |
BAA21499 | |
CLEC11A | |
E. coli-derived recombinant human SCGF-beta Gly19-Phe245 Accession # BAA21499 | |
100 μg | |
Primary | |
Detects human SCGF/CLEC11a in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CLEC11a, CLECSF3, LSLCL, P47, SCGF | |
Sheep | |
Affinity Purified | |
RUO | |
6320 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only